Learn More
BD Taxo™ Anaerobe Disc Set
Catalog No. B31651 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
5 Cartridges
Catalog No. Quantity
B31651 5 Cartridges
1 options
This item is not returnable. View return policy
Catalog No. B31651 Supplier BD Supplier No. 231651
Add to Cart
Add to Cart
This item is not returnable. View return policy
For presumptive identification of Gram-negative anaerobic Bacilli
  • Furnished in cartridges containing 50 discs each
  • Each disc is prepared from absorbent paper impregnated with antimicrobial agents
  • Last disc in each cartridge is marked “X” and contains drug as coded
  • Cartridges are for use in BD BBL™ single disc dispensers
  • Each cartridge includes five antimicrobial agents
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Colistin, Kanamycin, Penicillin, Rifampin, Vancomycin
Content And Storage -20°C to +8°C
Includes 1mg Kanamycin, 15μg Rifampin, 2U Penicillin, 5μg Vancomycin and 10μg Colistin
Packaging Type Disc
Quantity 5 Cartridges