Learn More
BD Taxo™ Differentiation Discs: Antibiotic - Taxo SPS
Catalog No. B4331726 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1 Pack
Catalog No. Quantity
B4331726 1 Pack
1 options
This item is not returnable. View return policy
Catalog No. B4331726 Supplier BD Supplier No. 231726
 Restricted Item
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Detect, differentiate and identify microorganisms
  • Organisms react with chemical agents impregnated on discs resulting in positive or negative reactions
  • Combination of positive and negative reaction aids in the identification of the organism
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents SPS
Content And Storage 2°C to 8°C
Packaging Type Disc
Quantity 1 Pack
For in vitro Diagnostic use. Follow established laboratory procedures in handling and disposing of infectious materials.