Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Tubed Media Letheen Broth

Catalog No. L98239 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L98239 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L98239 Supplier BD Supplier No. 298239
Add to Cart
Add to Cart

For microbiological testing of cosmetics

TRUSTED_SUSTAINABILITY

Specifications

Broth Type Letheen Broth
Certifications/Compliance ISO/DIS, 11014
Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Packaging Type Tube
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.