Learn More
Medex Supply BD ULTRA-FINE III SHRT PEN 31G
SDP
Catalog No. NC1612920
Encompass_Preferred
BD Ultra-Fine III Short Pen Needle, 31 G x 5/16"(8MM), 100/bx; BD Model: 320109
This item is not returnable. View return policy
Catalog No. NC1612920 Supplier Medex Supply Supplier No. BD320109
BD Ultra-Fine III Short Pen Needle, 31 G x 5/16"(8MM), 100/bx; BD Model: 320109