Learn More
BD Universal Viral Transport Collection Kits
Catalog No. B220529 Shop All Waters Corporation Products
Change view
Click to view available options
Swab Configuration:
Dual Applicator (Flex, Regular)
Single Applicator
Single Applicator (Minitip)
Single Flexible Applicator
Tip Type:
Minitip Flocked
Regular Flocked
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Swab Configuration Tip Type
B220529 Single Applicator Minitip Flocked
B220528 Single Applicator Regular Flocked
B220531 Single Applicator (Minitip) Minitip Flocked
B220526 Single Flexible Applicator Minitip Flocked
B220527 Dual Applicator (Flex, Regular) Regular Flocked
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. B220529 Supplier BD Supplier No. 220529
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Sterile kits provide room temperature (2° to 25°C) storage and transport of viruses, chlamydiae, mycoplasmas and ureaplasmas
For the collection and transport of clinical specimens containing viruses, chlamydiae, mycoplasmas or ureaplasmas from the collection site to the testing laboratory. System can be processed using standard clinical laboratory operating procedures for viral, chlamydial, mycoplasmal and ureaplasmal culture. Includes a universal transporting medium that is room temperature stable, which can sustain viability (and infectivity) of a plurality of organisms during transit to the testing laboratory. Medium includes protein for stabilization, antibiotics to minimize bacterial and fungal contamination, and a buffer to maintain a neutral pH.
  • Need for refrigerated storage and transport on ice is eliminated, decreasing the prevalence of specimen rejection and eliminating messy specimen transport
  • Feature a unique blend of antimicrobics that target and suppress bacterial and fungal contamination and are not harmful to viruses, chlamydiae, mycoplasmas or ureaplasmas
  • Clinical pathogens are maintained, while growth of contaminants is inhibited
  • Preserves viruses and chlamydiae when stored frozen with the presence of a cryoprotectant
  • Provided with labeled capture-cap vials designed for transport of the clinical sample
  • System is also supplied as a sample collection kit that comprises a package containing one capture-cap vial of medium and a peel pouch incorporating sterile polyester or nylon flocked specimen collection swabs, with scored shafts for easy breakage
  • Capture-cap is designed to secure the shaft of the swab sample to the cap, eliminating use of forceps to remove swab in the laboratory
  • TRUSTED_SUSTAINABILITY
    Specifications
    Product Type Viral Transport Collection Kit
    Sterility Sterile
    Swab Configuration Single Applicator
    For Use With (Application) Collection and transport of clinical specimens containing Viruses, Chlamydiae, Mycoplasmas or Ureaplasmas from the collection site to the testing laboratory
    Includes 3mL Vial, one sterile Nylon flocked tip swab, scored Plastic shaft
    Quantity 50 Pk.
    Shaft Material Plastic
    Tip Material Nylon
    Tip Type Minitip Flocked
    Volume (Metric) 3 mL