Learn More
Description
- Streptavidin protein conjugated to BD Horizon V500
- Clone: Streptavidin
- Concentration: 0.5mg/mL
- Reactivity: QC Testing: Mouse; Tested in development: Human
Flow Cytometry
Specifications
Specifications
| Concentration | 0.5mg/mL |
| Cross Reactivity | Mouse |
| pH | 7.3 |
| Quantity | 0.1 mg |
| Content And Storage | 4°C |
| Form | Solution |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse Streptavidin BD Horizon V500 |
Safety and Handling
For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.