Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Vacutainer™ Blood Collection Needles

Catalog No. p-178295
Click to view available options
Gauge:
1 in.
Length (English):
21 in.
22 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Sterile, disposable, double-ended needles

  • May be affixed to the BD Vacutainer Needle Holder, (available separately) with minimum danger of contamination
  • Multiple sample needles feature a dry, natural rubber sleeve that covers the opening on the stopper-puncturing end of the needle and stops flow of blood as evacuated tube is removed
  • Color-coded plastic hub and needle case provide easy gauge identification

Order Info

Packaged in plastic hardpack with tamper-evident label

TRUSTED_SUSTAINABILITY

Specifications

Gauge 1 in.
Length (English) 21 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.