Learn More
BD Vacutainer™ General Use Syringe Needles
Catalog No. 23021020 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Needle Gauge:
18 G
Catalog No. Needle Gauge
23-021-020 18 G
1 options
This item is not returnable. View return policy
Catalog No. 23-021-020 Supplier BD Supplier No. 305180
Add to Cart
Add to Cart
This item is not returnable. View return policy
Blunt fill needle provides a substitute for conventional needles
  • Provides the convenience of pre-attached combos along with clinical flexibility
  • Bridges medication preparation and administration; effective devices for filling and transferring medications from vials, ampoules and IV bags to their final medication delivery reservoirs
  • Designed to minimize the risk of needlesticks; requires 10X more puncture penetration force than a conventional sharp
  • Lower cost alternative than sharps safety engineered devices
  • 18 gauge blunt needles allow for rapid transfer and aspiration without coring
  • 1.5 in. can easily penetrate both single and double septum barriers
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 18 G
For Use With (Application) Provides a substitute for conventional needles, general use
Length (English) Needle 1.5 in.
Product Type Syringe Needle
Sterility Sterile