Learn More
BD Vacutainer™ Glass Blood Collection Tubes with Fluoride: Hemogard™
Catalog No. 0268869 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Capacity (Metric):
7 mL
Closure Color:
Gray
Catalog No. Capacity (Metric) Closure Color
02-688-69 7 mL Gray
1 options
This item is not returnable. View return policy
Catalog No. 02-688-69 Supplier BD Supplier No. 367729
Add to Cart
Add to Cart
This item is not returnable. View return policy
Evacuated glass blood collection tubes for glucose determinations
  • BD Vacutainer™ fluoride tubes are used to collect samples for glucose determinations
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are gray
Hemogard™ Stoppers
  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube
TRUSTED_SUSTAINABILITY
Specifications
Additive Sodium Fluoride (30 mg)
Closure Type Hemogard
Capacity (Metric) 7 mL
Closure Color Gray