Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Vacutainer™ Glass Serum Tubes with Uncoated Interior

Catalog No. 0268362 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Capacity (Metric):
7 mL
Closure Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
02-683-62 7 mL Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-683-62 Supplier BD Supplier No. 366442

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Evacuated glass serum tubes for general blood sample collection

  • Tube interiors are sterile
  • Tube stoppers are lubricated with glycerin or silicone to facilitate stopper insertion

  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are red for easy identification

TRUSTED_SUSTAINABILITY

Specifications

Color Red Cap
Additive No Additive
Material Glass
Sterility Sterile
Closure Type Traditional Cap
Capacity (Metric) 7 mL
Closure Color Red
Label Type Paper
Length (Metric) 100 mm
Diameter (Metric) Outer 13 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.