Learn More
BD Vacutainer™ Glass STAT Chemistry Tubes
Catalog No. 0268390 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Tube Size:
16 x 100mm
Catalog No. Tube Size
02-683-90 16 x 100mm
1 options
This item is not returnable. View return policy
Catalog No. 02-683-90 Supplier BD Supplier No. 366525
This item is not returnable. View return policy
Evacuated glass blood collection tubes for stat chemistry determinations
  • BD Vacutainer serum tubes with Thrombin additive are used for stat chemistry determinations
  • Serum tubes are coated with silicone and micronized silica particles to accelerate clotting; silicone coating on the walls of most serum tubes reduces adherence of red cells to tube walls
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Hemogard™ Closures
  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube
TRUSTED_SUSTAINABILITY
Specifications
Tube Size 16 x 100mm