Learn More
Description
Needle heel is specially manufactured to minimize catheter shearing
- Modified Tuohy point is designed to provide enhanced tactile feel
- Unique one-piece plastic style is fitted to prevent tissue coloring and is lighter to enhance tactile feel during insertion
- Calibrated needle depth markings assist in awareness of needle placement depth
- Needle length: 127mm (5 in.)
- Needle gauge: 17 G
- Self-life: 1825 days
- Modified point
- Fixed wings
- Single-use
- Thin wall
- Sterile
Specifications
Specifications
| Product Type | Epidural Needle |
| Needle Gauge | 17 G |
| Length (English) Needle | 5 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.