Learn More
BD Vacutainer™ Plastic Blood Collection Tubes No Additives: Bulk Pack
Catalog No. 23021630 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Additive:
No Additive
Catalog No. Additive
23-021-630 No Additive
1 options
This item is not returnable. View return policy
Catalog No. 23-021-630 Supplier BD Supplier No. 367982
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Plastic tubes with no additives for venous blood and urine collection
  • Blood is drawn directly from the vein into the evacuated collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Made of polyethylene terephthalate (PET)
  • Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; and (2) eliminating breakage when tubes are centrifuged at higher speeds 
  • In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation
  • For use with Hemogard™ Closures, which provide a safety shield for safer tube handling
 Note:  Closures are not included; sold separately.
Order Info
Packaged in bulk bag
TRUSTED_SUSTAINABILITY
Specifications
Additive No Additive
Material Plastic
Sterility Non-sterile
Closure Type Rubber Stopper
Tube Size 16 x 100 mm