Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD SafetyGlide™ Insulin, TB and Allergy Syringe with Needle

Catalog No. 1368914
Change view
Click to view available options
Needle Gauge:
31 G
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
13-689-14 31 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-689-14 Supplier BD Supplier No. 305937
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The permanently attached needle is designed with low waste space, which may result in reduced medication waste

Tiny Needle Technology completes the BD SafetyGlide line to meet your Sub-Q and intradermal injection need. Includes safety shield.

Specifications

Needle Gauge 31 G
Length (English) Needle 0.312 in.
Product Type Insulin Syringe
Volume (Metric) 0.3 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.