Learn More
BD Vacutainer™ Urine Culture and Sensitivity (C&S) Transport Kits
Catalog No. 14375131 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Volume (Metric):
5 mL
Catalog No. Volume (Metric)
14-375-131 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 14-375-131 Supplier BD Supplier No. 364949
This item is not returnable. View return policy
Contain a freeze-dried preservative for transporting and preserving urine specimens
  • The kit comes with a BD Vacutainer tube and a transfer straw that delivers urine from the cup into the evacuated tube
  • Preservative prevents overgrowth of bacteria, even if the specimen is contaminated
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 5 mL