Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Plastic Serum Tubes, Silicone-Coated, with Conventional Stopper

Catalog No. 02685112 Shop All Waters Corporation Products
Change view
Click to view available options
Diameter (Metric) Outer:
13 x 75 mm
13 x 100 mm
16 x 100 mm
Capacity (Metric):
3 mL
6 mL
10 mL
Product Type:
Barcoded Blood Collection Tube
Blood Collection Tube
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Capacity (Metric) Product Type
02-685-112 16 x 100 mm 10 mL Blood Collection Tube
02-657-27 13 x 75 mm 3 mL Blood Collection Tube
23-021-018 16 x 100 mm 10 mL Barcoded Blood Collection Tube
23-021-012 13 x 100 mm 6 mL Blood Collection Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 02-685-112 Supplier BD Supplier No. 367820
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for serum determinations in chemistry, serology and immunohematology.

  • Sterile vacuum blood collection tubes for serum determination
  • Spray-coated silica interior
  • Tubes marked as Best Practice Formulary meet demanding requirements of complex diagnostic instruments and current clinical guidelines
  • With conventional tube stopper

Evacuated Tubes Simplify Blood Collection

  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Plastic Evacuated Tubes
  • Made of polyethylene terephthalate (PET)
  • Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
  • In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation

TRUSTED_SUSTAINABILITY

Specifications

Product Type Blood Collection Tube
Additive Clot Activator, Silicone Coated
Material Plastic
Closure Type Traditional Cap
Capacity (Metric) 10 mL
Closure Color Red
Diameter (Metric) Outer 16 x 100 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.