Learn More
BD Vacutainer™ Winged Blood Collection Sets
Catalog No. 0266539 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Needle Gauge:
23 G
25 G
Length (English) Tube:
7 in.
12 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (English) Tube
02-665-39 23 G 12 in.
02-665-43 23 G 7 in.
02-665-41 25 G 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 02-665-39 Supplier BD Supplier No. 367253
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
For performing difficult draws from veins directly into a BD Vacutainer evacuated tube
  • Shorter (0.75 in. [1.9cm]), smaller-gauge needles glide smoothly into the vein with little discomfort to the patient
  • Flexible wings are color coded according to needle gauge
  • Closed system, with multisample Luer adapter (MSLA), facilitates multiple-tube collections
  • Eliminate spillage as well as difficult, time-consuming syringe-tube manipulations
  • Ideal for individuals whose veins make straight forward collection difficult
TRUSTED_SUSTAINABILITY
Specifications
Product Type Blood Collection Set
Needle Gauge 23 G
Length (English) Needle 0.75 in.
Length (English) Tube 12 in.