Learn More
BD Vacutainer™ Winged Blood Collection Sets
Catalog No. 0266541 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Needle Gauge:
23 G
25 G
Length (English) Tube:
7 in.
12 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (English) Tube
02-665-41 25 G 12 in.
02-665-39 23 G 12 in.
02-665-43 23 G 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 02-665-41 Supplier BD Supplier No. 367255
This item is not returnable. View return policy
For performing difficult draws from veins directly into a BD Vacutainer evacuated tube
  • Shorter (0.75 in. [1.9cm]), smaller-gauge needles glide smoothly into the vein with little discomfort to the patient
  • Flexible wings are color coded according to needle gauge
  • Closed system, with multisample Luer adapter (MSLA), facilitates multiple-tube collections
  • Eliminate spillage as well as difficult, time-consuming syringe-tube manipulations
  • Ideal for individuals whose veins make straight forward collection difficult
TRUSTED_SUSTAINABILITY
Specifications
Product Type Blood Collection Set
Needle Gauge 25 G
Length (English) Needle 0.75 in.
Length (English) Tube 12 in.