Learn More
BD Vacutainer™ Winged Safety Push Button Blood Collection Sets without Luer Adapter
Catalog No. BD367324 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Needle Gauge:
23 G
25 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
BD367324 23 G
BD367323 25 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BD367324 Supplier BD Supplier No. 367324
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Blood collection set offers in-vein safety activation provides split-second protection against accidental needlestick injuries
  • Ease of use reduces training requirements and improves activation compliance
  • In-vein activation reduces the risk exposure to a contaminated needle
  • Easy activation without additional patient discomfort
  • Ideal for use in high-risk environments No luer adapter
TRUSTED_SUSTAINABILITY
Specifications
Product Type Push Button Blood Collection Set
Needle Gauge 23 G
Length (English) Needle 3/4 in.
Length (English) Tube 12 in.