Learn More
Med Vet International BD Vacutainer Plus Venous Blood Collection Tube, Sodium Citrate Additive, 2.7 mL, Hemogard™ Closure Plastic Tube, 100/Box
Catalog No. 503043227
Encompass_Preferred
Features Buffered Sodium Citrate, 0.109M (Equiv. to 3.2% Sodium Citrate)
Catalog No. 50-304-3227 Supplier Med Vet International Supplier No. 363083
Features Buffered Sodium Citrate, 0.109M (Equiv. to 3.2% Sodium Citrate)