Learn More
BD Veritor™ System RSV Control Swab Set
Catalog No. B256061 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 Pairs
Catalog No. Quantity
B256061 10 Pairs
1 options
This item is not returnable. View return policy
Catalog No. B256061 Supplier BD Supplier No. 256061
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Each pair include one positive control swab (RSV antigen [noninfectious cell lysate]) with <0.1% sodium azide (preservative) and one negative control swab (detergent-treated non-infected cells) with <0.1% sodium azide (preservative).
Ten pairs of swabs
TRUSTED_SUSTAINABILITY
Specifications
Description RSV Control Swab Set
For Use With (Application) For Rapid Detection of Respiratory Syncytial Virus (RSV)
Includes Each pair include one positive control swab (RSV antigen [noninfectious cell lysate]) with <0.1% sodium azide (preservative) and one negative control swab (detergent-treated non-infected cells) with <0.1% sodium azide (preservative).
Quantity 10 Pairs
For in vitro diagnostic use only.