Learn More
BD Veritor™ System Flu A+B Control Swab Set
Catalog No. 22351287 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
22-351-287 Each
1 options
This item is not returnable. View return policy
Catalog No. 22-351-287 Supplier BD Supplier No. 256051
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
For Rapid Detection of Flu A+B
  • Easy sample processing - Prefilled unitized tube facilitates workflow
  • Ready in minutes - Test device is ready to insert into reader 10 minutes after sample is added
  • Insert and read - Simple one-touch button readies the reader for test device insertion
  • Objective results - Once the test device is inserted in the reader, an objective, digital test result is displayed in 10 seconds
TRUSTED_SUSTAINABILITY
Specifications
Description Veritor Control Swab Flu A&B 10 pr.
Clia Complexity Waived
Detectable Analytes Influenza A and B
Type Control Swab Set
For Use With (Application) Detection of Flu A + B
Quantity Each