Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Veritor™ System Clinical Flu A+B Kit

Catalog No. B256045 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
30 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B256045 30 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B256045 Supplier BD Supplier No. 256045
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

For Rapid Detection of Flu A+B

  • Streamlined workflow – Requires minimal hands-on time
  • Ready in minutes - Test device is ready to insert in reader 5 to 10 minutes after sample is added, depending on the assay
  • Insert and read - Simple one-touch button readies the BD Veritor Plus Analyzer for test device insertion
TRUSTED_SUSTAINABILITY

Specifications

Description Flu A + B Test Kit (For use in point of care settings)
Clia Complexity Waived
Detectable Analytes Influenza A and B
Sample Type Nasal swabs and nasopharyngeal swabs
Type Flu A + B Test Kit
For Use With (Application) Detection of Flu A + B
Includes Lateral flow test strips, pipettes, Flu A+/B- control swab, Flu B+/A- control swab, RV reagent C in tubes
Packaging Type 1 Kit
Quantity 30 Assays
Test Time 10 min.

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.