Learn More
Selleck Chemical LLC BD750 S0981-5MG
SDP
Catalog No. 5000041715
Encompass_Preferred
BD750 is an immunosuppressant and a dual inhibitor of JAK3 and STAT5 that inhibits IL-2-induced JAK3/STAT5-dependent T cell proliferation with IC50 of 1 5 M and 1 1 M for mouse and human T-cell proliferation respectively
Catalog No. 50-000-41715 Supplier Selleck Chemical LLC Supplier No. S09815MG
BD750 is an immunosuppressant and a dual inhibitor of JAK3 and STAT5 that inhibits IL-2-induced JAK3/STAT5-dependent T cell proliferation with IC50 of 1 5 M and 1 1 M for mouse and human T-cell proliferation respectively