Learn More
ATCC BDCM; Acute Myeloid Leukemia; Human (Homo sapiens)
Catalog No. 502384841
Encompass
BDCM is a B lymphoblast cell line that was isolated from the peripheral blood of a male patient with acute myelogenous leukemia. This cell line was deposited by RA Steinman and can be used in immunology and immune system disorder research.
Catalog No. 50-238-4841 Supplier ATCC Supplier No. CRL2740
BDCM is a B lymphoblast cell line that was isolated from the peripheral blood of a male patient with acute myelogenous leukemia. This cell line was deposited by RA Steinman and can be used in immunology and immune system disorder research.