Learn More
BDCS, silylation reagent, AcroSeal™, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | Yellow |
| Density | 0.9590g/mL |
| Flash Point | 58°C |
| Infrared Spectrum | Authentic |
| Packaging | AcroSeal™ Glass bottle |
| Refractive Index | 1.4400 to 1.4420 |
| Quantity | 100 mL |
| Fieser | 07,184; 01,489; 03,157; 08,255 |
| Specific Gravity | 0.959 |
| Formula Weight | 218.8 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H StatementFlammable liquid and vapour.Harmful if swallowed.Causes severe skin burns and eye damage.Suspected of damaging the unborn child.Contact with water liberates toxic gas. |
| Precautionary Statement | GHS P StatementKeep away from heat/sparks/open flames/hot surfaces.- No smoking.Wear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes.Remove conta |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.