Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDE No 153 Solution, 50 μg/mL in isooctane, MilliporeSigma™ Supelco™

Catalog No. 111018131 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8131 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8131 Supplier MilliporeSigma Supelco Supplier No. 336831ML
Add to Cart
Add to Cart

BDE No 153 is a contaminant, which can also be commercially used as a flame retardant additive in building materials, textiles, plastics, and electronic equipment.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 68631-49-2
Flash Point -12°C
Linear Formula C12H4 Br6 O
MDL Number MFCD06407722
Synonym 2,2 ′,4,4 ′,5,5 ′-Hexabromodiphenyl ether solution; 2,2 ′,4,4 ′,5,5 ′-HexaBDE; PBDE 153
Type Analytical Standard
Quantity 1 mL
Formula Weight 643.58
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 50 μg/mL in isooctane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.