Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDE No 205 Solution, 50 μg/mL in isooctane, MilliporeSigma™ Supelco™

Catalog No. 111018103 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8103 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8103 Supplier MilliporeSigma Supelco Supplier No. 336881ML
Add to Cart
Add to Cart

BDE No 205 is a pollutant belonging to the class of polybrominated diphenyl ethers. It can be commercially used as a flame retardant additive in building materials, textiles, plastics and electronic equipment.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 446255-56-7
Flash Point -12°C
Linear Formula C12H2 Br8 O
MDL Number MFCD09037729
Synonym 2,3,3 ′,4,4 ′,5,5 ′,6-Octabromodiphenyl ether solution; 2,3,3 ′,4,4 ′,5,5 ′,6-OctaBDE; PBDE 205
Type Analytical Standard
Quantity 1 mL
Formula Weight 801.38
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 50 μg/mL in isooctane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.