Learn More
ATCC Bdellovibrio bacteriovorus; Strain 100
Supplier:  ATCC 15356
Encompass
Bdellovibrio bacteriovorus strain 100 [NCIB 9529] is a whole-genome sequenced type strain that was isolated from soil in California. This bacterium is distributed in the plant pathogen Erwinia amylovora (ATCC 15357).
Catalog No. 50-238-4334