Learn More
Andwin Scientific Ethanol is composed of 63-64% ethanol and 29-31% water 4L
SDP
Catalog No. NC0678619
Encompass_Preferred
Ethanol is composed of 63-64% ethanol and 29-31% water. It has a Health Hazard NFPA Rating of 1, a Fire Hazard NFPA Rating of 3, and a Reactivity NFPA Rating of 0. It is packaged and sold in a 4 liter poly bottle.
Catalog No. NC0678619 Supplier Andwin Scientific Supplier No. BDH11564LP
Ethanol is composed of 63-64% ethanol and 29-31% water. It has a Health Hazard NFPA Rating of 1, a Fire Hazard NFPA Rating of 3, and a Reactivity NFPA Rating of 0. It is packaged and sold in a 4 liter poly bottle.