Learn More
Andwin Scientific BDH Alcohol 70%, 4x4L
SDP
Catalog No. NC0251360
Encompass_Preferred
BDH Alcohol 70% 4L Poly BDH 70% Alcohol A Blend Of 70% Reagent Alcohol (Denatured) And 30% Water Is Suitable For General Laboratory Use Including Cytology And Histology.
Catalog No. NC0251360 Supplier Andwin Scientific Supplier No. BDH11644LP
BDH Alcohol 70% 4L Poly BDH 70% Alcohol A Blend Of 70% Reagent Alcohol (Denatured) And 30% Water Is Suitable For General Laboratory Use Including Cytology And Histology.