Learn More
Andwin Scientific BDH ALCOHOL, 90% 4L 4/CASE
SDP
Catalog No. NC0453684
Encompass_Preferred
Ethanol is a clear colorless liquid chemical that has a mild alcohol-like odor. It is composed of 60-85% ethanol, 10-30% water, 3-5% isopropanol, and 2-5% methanol and is made to be used as a solvent. This product is classified as toxic. It is packaged in a 4 L poly bottle and is sold in a case of 4.
Catalog No. NC0453684 Supplier Andwin Scientific Supplier No. BDH11604LP
Ethanol is a clear colorless liquid chemical that has a mild alcohol-like odor. It is composed of 60-85% ethanol, 10-30% water, 3-5% isopropanol, and 2-5% methanol and is made to be used as a solvent. This product is classified as toxic. It is packaged in a 4 L poly bottle and is sold in a case of 4.