Learn More
Andwin Scientific BDH ARSTR WTR /LW TOC 1L 6CS
SDP
Catalog No. NC0643276
Encompass_Preferred
The water with low TOC can be used for analytical and preparative separations, gradient and isocratic conditions, and LC, HPLC, and trace organic analysis. This product is packaged in a 1-liter amber glass bottle and is sold in a case of 6.
This item is not returnable. View return policy
Catalog No. NC0643276 Supplier Andwin Scientific Supplier No. 87003650
The water with low TOC can be used for analytical and preparative separations, gradient and isocratic conditions, and LC, HPLC, and trace organic analysis. This product is packaged in a 1-liter amber glass bottle and is sold in a case of 6.