Learn More
Andwin Scientific Buffer Reference Standard pH 1.68 +- 0.02 Colorless 500 mL
SDP
Catalog No. NC0097206
Encompass_Preferred
The BDH pH reference standard buffer is a clear, colorless, odorless, liquid solution. This contains potassium tetroxalate dihydrate and deionized water. It is classified to have a 1.68 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.
Catalog No. NC0097206 Supplier Andwin Scientific Supplier No. BDH5006500ML
The BDH pH reference standard buffer is a clear, colorless, odorless, liquid solution. This contains potassium tetroxalate dihydrate and deionized water. It is classified to have a 1.68 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.