Learn More
Andwin Scientific BDH BUFFER PH 10 BLUE 500ML
SDP
Catalog No. NC0097212
Encompass_Preferred
The BDH pH reference standard buffer is a clear, blue, odorless, liquid solution. It is classified to have a 9.99-10.01 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.
Catalog No. NC0097212 Supplier Andwin Scientific Supplier No. BDH5072500ML
The BDH pH reference standard buffer is a clear, blue, odorless, liquid solution. It is classified to have a 9.99-10.01 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.