Learn More
Andwin Scientific BDH NH4OH ACS 28-30% 2.5L
SDP
Catalog No. NC9011254
Encompass_Preferred
Ammonium Hydroxide is a clear colorless liquid chemical that has an intense, suffocating odor of ammonia. With a pH greater than 11, this chemical is classified as a base. It is also categorized as hazardous. It is packaged in a 2.5 L glass bottle.
Catalog No. NC9011254 Supplier Andwin Scientific Supplier No. BDH30162.5LG
Ammonium Hydroxide is a clear colorless liquid chemical that has an intense, suffocating odor of ammonia. With a pH greater than 11, this chemical is classified as a base. It is also categorized as hazardous. It is packaged in a 2.5 L glass bottle.