Learn More
Andwin Scientific BDH NITRIC ACID ULTRA 500ML
SDP
Catalog No. NC0466600
Encompass
Nitric acid is a clear, colorless to yellow liquid chemical that has a strong acidic odor. It is composed of 60-70% nitric acid and water as a balance. This chemical is made to be used for quantitative trace metal analysis at the parts-per-trillion (ppt) level. Nitric acid is classified as hazardous and flammable. It is packaged in a 500 mL FEP bottle and is sold individually.
Catalog No. NC0466600 Supplier Andwin Scientific Supplier No. 87003226
Nitric acid is a clear, colorless to yellow liquid chemical that has a strong acidic odor. It is composed of 60-70% nitric acid and water as a balance. This chemical is made to be used for quantitative trace metal analysis at the parts-per-trillion (ppt) level. Nitric acid is classified as hazardous and flammable. It is packaged in a 500 mL FEP bottle and is sold individually.