Learn More
Andwin Scientific pH Test Strips, 0-14 range
SDP
Catalog No. NC1087294
Encompass_Preferred
Used for rapid pH testing directly at the point of interest, strips are always ready for immediate use. Designed for safe testing of dangerous, poisonous, or aggressive liquids. Long handle protects user from contact with sample. Fixed dye cannot bleed into the sample, protecting it against contamination and enabling measurements even in weakly buffered or strongly alkaline solutions.
Catalog No. NC1087294 Supplier Andwin Scientific Supplier No. BDH35309.606
Used for rapid pH testing directly at the point of interest, strips are always ready for immediate use. Designed for safe testing of dangerous, poisonous, or aggressive liquids. Long handle protects user from contact with sample. Fixed dye cannot bleed into the sample, protecting it against contamination and enabling measurements even in weakly buffered or strongly alkaline solutions.