Learn More
Andwin Scientific BDH POT CHLOR, SATUR SOL 500ML
SDP
Supplier:  Andwin Scientific BDH72422
Encompass_Preferred
The Potassium Chloride is a saturated aqueous solution that is made to be used as a general chemical reagent. It is a clear liquid that is utilized to refill calomel reference electrodes and to adjust the ionic strength for calcium ion selective electrode. This product is sold in a 500 mL poly bottle.
Catalog No. NC0530073