Learn More
Aviva Systems Biology Corp BDH2 Antibody - middle region (ARP57360_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57360_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BDH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BDH2. Alternate names: DHRS6, EFA6R, FLJ13261, PRO20933, UCPA-OR, UNQ6308, SDR15C1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54019