Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences BDMA, Benzyldimethylamine 25 ML
SDP

Catalog No. 50980202
Encompass_Preferred

(N-Benzyl-N, N-Dimethylamine)
CAS #103-83-3 C₆H₅CH₂N(CH₃)₂ F.W. 135.21 g/mol
Boiling point: 177-180 °C

N-Benzyl-N, N-Dimethylamine is an amine accelerator for polymerization of epoxy resins. A direct alternative to DMP-30 with a longer shelf life and better penetration.

Catalog No. 50-980-202 Supplier Electron Microscopy Sciences Supplier No. 1140025
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(N-Benzyl-N, N-Dimethylamine)
CAS #103-83-3 C₆H₅CH₂N(CH₃)₂ F.W. 135.21 g/mol
Boiling point: 177-180 °C

N-Benzyl-N, N-Dimethylamine is an amine accelerator for polymerization of epoxy resins. A direct alternative to DMP-30 with a longer shelf life and better penetration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.