Learn More
Enzo Life Sciences Brain derived neurotrophic factor (human), (recombinant) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5460005
Encompass_Preferred
Alternative name: BDNF. Purity: ≥97%. Formulation: Lyophilized from a 0.2µm sterile water solution. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-2082