Learn More
Medchemexpress LLC HY-P7116A 10ug , BDNF, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP7116A10UG
Encompass_Preferred
HY-P7116A 10ug BDNF, Human CAS: BDNF Protein, Human is a neurotrophin binding to the high-affinity tropomyosin-related receptor kinase B (TrkB) receptor to regulate neurodevelopmental processes, including neuronal survival, neuronal differentiation, and synaptic plasticity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1587