Learn More
Medchemexpress LLC BDNF Human CHO 25ug
SDP
Catalog No. 5000196466
Encompass_Preferred
Recombinant human brain-derived neurotrophic factor (BDNF) produced in CHO cells and supplied lyophilized for research use. This tag-free, glycosylated protein binds the TrkB receptor and is used to study neuronal survival, differentiation, and synaptic plasticity in cell-based assays and biochemical experiments.
  • Tag-free, CHO-expressed protein with native-like glycosylation.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) suitable for cell-based assays.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Available in small vial sizes for in vitro experimentation and dose optimization.
Catalog No. 50-001-96466 Supplier Medchemexpress LLC Supplier No. HYP711625UG
Recombinant human brain-derived neurotrophic factor (BDNF) produced in CHO cells and supplied lyophilized for research use. This tag-free, glycosylated protein binds the TrkB receptor and is used to study neuronal survival, differentiation, and synaptic plasticity in cell-based assays and biochemical experiments.
  • Tag-free, CHO-expressed protein with native-like glycosylation.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) suitable for cell-based assays.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Available in small vial sizes for in vitro experimentation and dose optimization.