Learn More
Broadpharm BDP 558/568 NHS ester, 25 mg
Catalog No. NC1833393
Encompass
BDP 558/568 is a borondipyrromethene fluorophore with emission in the yellow part of the spectrum. Its absorption and emission spectra are in similar range as TAMRA, BDP TMR, Cyanine3, and sulfo-Cyanine3. This is an amine reactive NHS ester.
Catalog No. NC1833393 Supplier Broadpharm Supplier No. BP2389425MG
BDP 558/568 is a borondipyrromethene fluorophore with emission in the yellow part of the spectrum. Its absorption and emission spectra are in similar range as TAMRA, BDP TMR, Cyanine3, and sulfo-Cyanine3. This is an amine reactive NHS ester.