Learn More
BD Cell Analysis 3P BDTM STEM CELL CTRL KT
Catalog No. 50620456
Encompass
Product shelf life is 6 weeks. Confirm product expiry date when ordering. Stem Cell Control Kit (Bi-Level Control). 1 x 2.0 ml CD34+ Low approx. 10 cells/ul, and 1 x 2.0 ml CD34+ High approx. 35 cells/ul.
Catalog No. 50-620-456 Supplier BD Cell Analysis 3P Supplier No. 340991
Product shelf life is 6 weeks. Confirm product expiry date when ordering. Stem Cell Control Kit (Bi-Level Control). 1 x 2.0 ml CD34+ Low approx. 10 cells/ul, and 1 x 2.0 ml CD34+ High approx. 35 cells/ul.