Learn More
Medchemexpress LLC HY-136789 5mg Medchemexpress, BDTX-189 CAS:2414572-47-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1367895MG
Encompass_Preferred
Medchemexpress, HY-136789 5mg BDTX-189 CAS:2414572-47-5 BDTX-189 is a potent, orally active and selective inhibitor of allosteric EGFR and HER2 oncogenic mutations, including EGFR/HER2 exon 20 insertion mutants. BDTX-189 shows KDs of 0.2, 0.76, 13 and 1.2 nM for EGFR, HER2, BLK and RIPK2, reapectively. Anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1460