Learn More
Electron Microscopy Sciences Beryllium Substrate Planchet, 101.6 mm x 1 mm
SDP
Catalog No. 5024683
Encompass_Preferred
These planchets are prepared from high purity beryllium (min. 98.5%) by electro-fusion to provide vacuum tight (~1x10-9atm-cm3/sec) and +/-10% dimensional tolerances.
Size: 101.6 mm x 1 mm
Catalog No. 50-246-83 Supplier Electron Microscopy Sciences Supplier No. 76017
These planchets are prepared from high purity beryllium (min. 98.5%) by electro-fusion to provide vacuum tight (~1x10-9atm-cm3/sec) and +/-10% dimensional tolerances.
Size: 101.6 mm x 1 mm