Learn More
ATCC BE(2)-M17; Neuroblastoma; Human (Homo sapiens)
Supplier:  ATCC CRL2267
Encompass
BE(2)-M17 is a neuroblast cell line that was isolated from the brain of a 2-year-old, male patient with neuroblastoma. This cell line was deposited by JL Biedler. It can be used in immunology, neuroscience and toxicology research.
Catalog No. 50-238-2098
 Restricted Item