Learn More
Chemglass Life Sciences 14L Bead Bath Only, 120V
SDP
Catalog No. 50143573
Encompass
14L Bead Bath Only, 120V. Bead bath gets up and running faster and you never need to turn it off. There is nothing to evaporate. It’s just like a trusty, always-on incubator. Unlike a standard bead-filled, water bath, Bead Bath allows you to achieve a new set temperature quickly, without long warm up delays, so you can make quick adjustments on the fly.With Bead Bath designed specifically for beads, temperatures are consistent and uniform everywhere in the bath. Thermal uniformity is an amazing ±0.5°C at 37°C and ±1.0 at 65°C.
Catalog No. 50-143-573 Supplier Chemglass Life Sciences Supplier No. CLS4598003
14L Bead Bath Only, 120V. Bead bath gets up and running faster and you never need to turn it off. There is nothing to evaporate. It’s just like a trusty, always-on incubator. Unlike a standard bead-filled, water bath, Bead Bath allows you to achieve a new set temperature quickly, without long warm up delays, so you can make quick adjustments on the fly.With Bead Bath designed specifically for beads, temperatures are consistent and uniform everywhere in the bath. Thermal uniformity is an amazing ±0.5°C at 37°C and ±1.0 at 65°C.